Learn More
Phospho-DARPP-32 (Ser137) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca:  Invitrogen PA535384
Detalles adicionales : Peso : 0.01000kg
Código de producto. 15388114
  • 473.00€ / 100 microlitros
Fecha estimada de envío: 23-04-2024
para ver el stock
Descripción
Descripción
This antibody is predicted to react with bovine, canine, human, mouse and non-human primate based on 100% sequence homology. This antibody contains enough material to conduct 10 mini-Western blots.
Phospho-DARPP-32 (Ser137) Polyclonal antibody specifically detects Phospho-DARPP-32 (Ser137) in Mouse, Rat samples. It is validated for Western Blot.
Especificaciones
Especificaciones
Phospho-DARPP-32 (Ser137)
Polyclonal
Unconjugated
Ppp1r1b
AU040756; DARPP32; DARPP-32; dopamine- and cAMP-regulated neuronal phosphoprotein; dopamine and cAMP-regulated neuronal phosphoprotein 32; dopamine- and cAMP-regulated phosphoprotein DARPP-32; FLJ20940; IPPD; neuronal phosphoprotein DARPP-32; OTTHUMP00000164275; OTTHUMP00000164276; Ppp1r1b; PPR1B; protein pho; protein phosphatase 1 regulatory inhibitor subunit 1B; protein phosphatase 1 regulatory subunit 1B; protein phosphatase 1, regulatory (inhibitor) subunit 1B
Rabbit
Affinity chromatography
RUO
19049, 360616
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
∽0.25 mg/mL
0.01M HEPES with 0.15M NaCl, 0.1mg/mL BSA, 50% glycerol and no preservative; pH 7.5
Q60829, Q6J4I0
Ppp1r1b
Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser137 of rat PPP1R1B conjugated to KLH
100 μL
Primary
Mouse, Rat