missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-Caspase 8 (Ser347) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA536709
Detalles adicionales : Peso : 0.00910kg
Descripción
This antibody detects endogenous protein at a molecular weight of 38 and 55 kDa. Purity is >95% by SDS-PAGE.
Phospho-Caspase 8 (Ser347) Polyclonal antibody specifically detects Phospho-Caspase 8 (Ser347) in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Especificaciones
Phospho-Caspase 8 (Ser347) | |
Polyclonal | |
Unconjugated | |
Casp8 | |
ALPS2B; apoptosis-related cysteine peptidase; apoptotic cysteine protease; Apoptotic protease Mch-5; CAP4; CASP8; CASP-8; caspase 8; caspase 8, apoptosis-related cysteine peptidase; caspase 8, apoptosis-related cysteine protease; Caspase8; caspase-8; Caspase-8 precursor; Caspase-8 subunit p10; Caspase-8 subunit p18; EC 3.4.22.61; FADD-homologous ICE/ced-3-like protease; FADD-like ICE; Fas-linked ICE-like protease; FLICE; FLJ17672; ICE8; ICE-like apoptotic protease 5; MACH; MACH-alpha-1/2/3 protein; MACH-beta-1/2/3/4 protein; MCH5; MGC78473; MORT1-associated ced-3 homolog; OTTHUMP00000163720; OTTHUMP00000165063; OTTHUMP00000206557; OTTHUMP00000206581 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
12370, 64044, 841 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.05% sodium azide; pH 7.2 | |
O89110, Q14790, Q9JHX4 | |
Casp8 | |
Synthetic phosphopeptide derived from human Caspase-8 around the phosphorylation site of Serine 347 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |