missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-beta Catenin (Tyr654) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA536748
Detalles adicionales : Peso : 0.00910kg
Descripción
This antibody detects endogenous protein at a molecular weight of 92 kDa. Purity is >95% by SDS-PAGE.
Phospho-beta Catenin (Tyr654) Polyclonal antibody specifically detects Phospho-beta Catenin (Tyr654) in Human, Mouse, Rat samples. It is validated for Western Blot.Especificaciones
Phospho-beta Catenin (Tyr654) | |
Polyclonal | |
Unconjugated | |
CTNNB1 | |
armadillo; armadillo homolog; Bcatenin; b-catenin; beta 1 88kDa; beta catenin; Betacatenin; beta-catenin; beta-catenin 1; Bfc; C20orf33; Cadherin associated protein; catenin; catenin (cadherin associated protein), beta 1; catenin (cadherin associated protein), beta 1, 88kDa; catenin (cadherin-associated protein) beta 1; catenin (cadherin-associated protein), beta 1; catenin (cadherin-associated protein), beta 1, 88kDa; Catenin b 1; Catenin b1; catenin beta; catenin beta 1; catenin beta 1 L homeolog; Catenin beta1; Catenin beta-1; Catenin β 1; Catenin β1; Catnb; CHBCAT; CTNB1; CTNNB; ctnnb1; ctnnb1.L; ctnnb1-a; ctnnb1-b; dJ633O20.1; DKFZp686D02253; FLJ25606; FLJ37923; id:ibd2058; Mesc; MRD19; NAP; NYD-SP19; OK/SW-cl.35; OTTHUMP00000209289; P14L; PP8304; PRO2286; RP5-1118M15.1; wu:fb73e10; wu:fi81c06; wu:fk25h01; XELAEV_18031149mg; β catenin; βcatenin | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
12387, 1499, 84353 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.05% sodium azide; pH 7.2 | |
P35222, Q02248, Q9WU82 | |
CTNNB1 | |
Synthetic phosphopeptide derived from human Catenin-beta around the phosphorylation site of Tyrosine 654 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |