missing translation for 'onlineSavingsMsg'
Learn More

ERK1/ERK2 Monoclonal Antibody (3F8B3), Invitrogen™

Mouse Monoclonal Antibody

Marca:  Invitrogen MA515605

Detalles adicionales : Peso : 0.01000kg

Código de producto. 11335573

  • 508.00€ / 100 microlitros
Fecha estimada de envío: 16-04-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

MA5-15605 targets p44/42 MAPK (Erk1/2) in FACS, IHC, WB and IF applications and shows reactivity with Human and mouse samples.

ERK1/ERK2 Monoclonal antibody specifically detects ERK1/ERK2 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry, Western Blot.
Especificaciones

Especificaciones

ERK1/ERK2
Monoclonal
1 mg/mL
PBS with 1mg/mL BSA and 0.05% sodium azide
P21708, P27361, P28482, P63085, P63086, Q63844
MAPK1, MAPK3
Purified recombinant fragment of human MAPK expressed in E. Coli.
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG2b
Immunocytochemistry, Immunohistochemistry, Western Blot
3F8B3
Unconjugated
MAPK3
12559; 9030612K14Rik; AA407128; AU018647; BcDNA:RE08694; C78273; CG12559; CG12559-PA; CG12559-PC; CG12559-PD; CG12559-PE; CG12559-PG; CG12559-PH; CG12559-PI; CG18732; CT34260; CT39192; DERK; D-ERK; DERK-A; Diphospho-ERK; dm-dpERK; Dmel\CG12559; Dmel_CG12559; DmErk; DmERKA; DmERK-A; DmMAPK; dpERK; dp-ERK; dpERK1; dpMAPK; drosophila ERK; Dsor2; E(sina)7; EC2-1; EK2-1; enhancer of seven in absentia 7; ERK; Erk MAP kinase; Erk/Map kinase; ERK/rolled; Erk1; Erk-1; erk1 erk2; Erk1/2; ERK1b; ERK2; ERK-2; ERKA; ERK-A; ERT1; ERT2; Esrk1; extracellular signal-regulated kinase; extracellular signal-regulated kinase (ERK2); extracellular signal-regulated kinase 1; Extracellular signal-regulated kinase 2; extracellular signal-regulated kinase A; extracellular signal-regulated kinase-1; extracellular signal-regulated kinase-2; extracellular signal-related kinase 1; Extracellular-regulated kinase A; extracellular-signal-regulated kinase 2; Extracellular-signal-related kinase A; EY2-2; fi06b09; GroupI
Mouse
Affinity chromatography
RUO
116590, 26413, 26417, 50689, 5594, 5595
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales