Learn More
p41-ARCb Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA528103
Detalles adicionales : Peso : 0.01000kg
Descripción
Recommended positive controls: NIH-3T3. Predicted reactivity: Mouse (100%), Rat (100%), Pig (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
p41-ARCb Polyclonal antibody specifically detects p41-ARCb in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Especificaciones
p41-ARCb | |
Polyclonal | |
Unconjugated | |
ARPC1B | |
41kDa; AA408064; AA408534; AA571392; actin related protein 2/3 complex subunit 1B; actin related protein 2/3 complex subunit 1B, 41kDa; actin related protein 2/3 complex, subunit 1B; actin related protein 2/3 complex, subunit 1B, 41kDa; actin-related protein 2/3 complex subunit 1B; Actin-related protein complex 1b; AF007010; ARC41; Arp2/3 complex 41 kDa subunit; ARP2/3 protein complex subunit p41; Arpc1b; AW208418; L72; OTTHUMP00000206020; p40-ARC; p41-ARC; SOP2Hs | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10095, 11867 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 10% glycerol and 0.01% thimerosal; pH 7 | |
O15143, Q9WV32 | |
ARPC1B | |
Synthetic peptide corresponding to a region within amino acids 1 and 44 of Human p41-ARCb | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |