missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LTB4R Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA528750
Detalles adicionales : Peso : 0.01000kg
Descripción
Recommended positive controls: 293T, H1299. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
LTB4R Polyclonal antibody specifically detects LTB4R in Human samples. It is validated for Western Blot.Especificaciones
LTB4R | |
Polyclonal | |
Unconjugated | |
LTB4R | |
BLT; BLT1; BLT-1; BLTR; Chemoattractant receptor-like 1; chemokine receptor-like 1; CMKRL1; G protein-coupled receptor 16; GPR16; G-protein coupled receptor 16; leukotriene B4 receptor; leukotriene B4 receptor 1; LTB4 receptor; Ltb4r; LTB4-R 1; Ltb4r1; LTB4-R1; LTBR1; mBLTR; P2RY7; P2Y purinoceptor 7; P2Y7; purinergic receptor P2Y, G-protein coupled, 7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1241 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q15722 | |
LTB4R | |
Synthetic peptide corresponding to a region within amino acids 1 and 45 of Human P2Y7 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |