missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P2Y6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA120617
Detalles adicionales : Peso : 0.01000kg
Descripción
PA1-20617 detects P2Y6 in human samples.
P2Y6 Polyclonal antibody specifically detects P2Y6 in Human samples. It is validated for Immunohistochemistry (Paraffin).Especificaciones
P2Y6 | |
Polyclonal | |
Unconjugated | |
P2ry6 | |
2010204J23Rik; G-coupled nucleotide receptor; MGC15335; P2 purinoceptor; P2ry6; P2Y ATP receptor 6; P2Y purinoceptor 6; p2y3 and chick; p2y3 receptor; P2Y6; P2Y6 receptor; PP2891; Purinoceptor p2y6; pyrimidinergic receptor P2Y, G-protein coupled, 6; pyrimidinergic receptor P2Y6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5031 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q15077 | |
P2ry6 | |
Synthetic peptide corresponding to the second cytoplasmic loop of human P2Y6. | |
25 μg | |
Primary | |
Human | |
Antibody | |
IgG |