missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P2X7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA528020
Detalles adicionales : Peso : 0.01000kg
Descripción
Predicted reactivity: Mouse (82%), Rat (81%), Dog (91%), Bovine (86%), Guinea pig (83%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
P2X7 Polyclonal antibody specifically detects P2X7 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin).Especificaciones
P2X7 | |
Polyclonal | |
Unconjugated | |
P2RX7 | |
AI467586; ATP receptor; MGC20089; P2rx7; P2X purinoceptor 7; p2X receptor 7; P2X(7); P2X7; P2X7 purinoceptor; P2X7 receptor; P2X7R; P2Z; P2Z receptor; Purinergic receptor; purinergic receptor P2X 7; purinergic receptor P2X, ligand gated ion channel, 7; purinergic receptor P2X, ligand-gated ion channel, 7; purinergic receptor P2X7; purinergic receptor P2X7 variant A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
18439, 29665, 5027 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q64663, Q99572, Q9Z1M0 | |
P2RX7 | |
Recombinant protein fragment corresponding to a region within amino acids 103 and 348 of Human P2X7 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |