missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P2Y8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA534098
Detalles adicionales : Peso : 0.01000kg
Descripción
Percent identity with other species by BLAST analysis: Human, Gorilla (100%) Marmoset (80%).
P2Y8 Polyclonal antibody specifically detects P2Y8 in Human samples. It is validated for Immunohistochemistry (Paraffin).Especificaciones
P2Y8 | |
Polyclonal | |
Unconjugated | |
P2RY8 | |
G-protein coupled purinergic receptor P2Y8; MGC50878; P2RY8; P2Y purinoceptor 8; P2Y8; purinergic receptor P2Y, G-protein coupled, 8; purinergic receptor P2Y8; RP11-261P4.4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
286530 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q86VZ1 | |
P2RY8 | |
Synthetic 15 amino acid peptide from 3rd cytoplasmic domain of human P2RY8 / P2Y8. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |