Learn More
p21 ARC Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marca: Invitrogen PA132486
Detalles adicionales : Peso : 0.01000kg
Descripción
Recommended positive controls: The peptide used to generate this antibody is available for purchase (GTX44635-PEP).. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
p21 ARC Polyclonal antibody specifically detects p21 ARC in Human, Mouse samples. It is validated for Western Blot.Especificaciones
p21 ARC | |
Polyclonal | |
Unconjugated | |
ARPC3 | |
1110006A04Rik; 21kDa; AA408672; actin related protein 2/3 complex subunit 3; actin related protein 2/3 complex subunit 3, 21kDa; actin related protein 2/3 complex, subunit 3; actin related protein 2/3 complex, subunit 3, 21kDa; actin-related protein 2/3 complex subunit 3; AI788639; ARC21; Arp2/3 complex 21 kDa subunit; Arp2/3 complex subunit p21-Arc; ARP2/3 protein complex subunit p21; Arpc3; p21-Ar; p21Arc; p21-Arc | |
Goat | |
Antigen affinity chromatography | |
RUO | |
10094, 56378 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
O15145, Q9JM76 | |
ARPC3 | |
synthetic peptide RQFMNKSLSGPGQ, corresponding to C terminal amino acids 165-177 of Human p21-ARC | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |