Learn More
RPRD1A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA530668
Detalles adicionales : Peso : 0.01000kg
Descripción
Recommended positive controls: HCT116. Predicted reactivity: Mouse (98%), Rat (98%), Rhesus Monkey (99%), Chimpanzee (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
RPRD1A Polyclonal antibody specifically detects RPRD1A in Human samples. It is validated for Immunocytochemistry, Western Blot.Especificaciones
RPRD1A | |
Polyclonal | |
Unconjugated | |
RPRD1A | |
C77387; CDKN2B-related protein; cyclin-dependent kinase 2B-inhibitor-related protein; cyclin-dependent kinase inhibitor 2B-related protein; Cyclin-dependent kinase inhibitor 2B-related protein (p15INK4B-related protein); HsT3101; Kiaa4077; mKIAA4077; p15INK4B-related protein; P15RS; regulation of nuclear pre-mRNA domain containing 1A; regulation of nuclear pre-mRNA domain-containing protein 1A; RGD1307374; Rprd1a | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55197 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.69 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q96P16 | |
RPRD1A | |
Recombinant fragment corresponding to a region within amino acids 103 and 312 of Human P15RS | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |