missing translation for 'onlineSavingsMsg'
Learn More

p130Cas Monoclonal Antibody (CAS-14 (35B-1A4)), Invitrogen™

Mouse Monoclonal Antibody

Marca:  Invitrogen MA514276

Detalles adicionales : Peso : 0.01000kg

Código de producto. 11355023

  • 473.00€ / 500 microlitros
Fecha estimada de envío: 23-04-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

MA5-14276 targets p130Cas in IHC (P), IP, and WB applications and shows reactivity with Human and Rat samples. The MA5-14276 immunogen is recombinant human p130Cas protein.

p130Cas Monoclonal antibody specifically detects p130Cas in Human, Rat samples. It is validated for Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.
Especificaciones

Especificaciones

p130Cas
Monoclonal
0.2 mg/mL
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
P56945, Q63767
BCAR1
Recombinant human p130Cas protein
500 μL
Primary
Human, Rat
Antibody
IgG1 κ
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
CAS-14 (35B-1A4)
Unconjugated
BCAR1
AI385681; BCA1; Bcar1; BCAR1 scaffold protein, Cas family member; BCAR1, Cas family scaffold protein; BCAR1, Cas family scaffolding protein; breast cancer anti-estrogen resistance 1; breast cancer anti-estrogen resistance protein 1; Cas; Cas scaffolding protein family member 1; CAS1; CASS1; CRKAS; CRK-associated substrate; Crk-associated substrate p130Cas; OTTHUMP00000174941; p130 Cas; P130CAS; p130Cas (pTyr410); p130Cas (pY410); p130Cas phospho; phospho p130Cas; v-crk-associated tyrosine kinase substrate
Mouse
Protein G
RUO
25414, 9564
4° C
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales