missing translation for 'onlineSavingsMsg'
Learn More

delta Catenin Monoclonal Antibody (6H11), Invitrogen™

Mouse Monoclonal Antibody

Marca:  Invitrogen 339700

Detalles adicionales : Peso : 0.50000kg

Código de producto. 10447173

  • 469.00€ / cada uno
Fecha estimada de envío: 16-04-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Description

Description

delta Catenin Monoclonal antibody specifically detects delta Catenin in Canine, Chicken, Human, Mouse, Rat samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.
Specifications

Specifications

delta Catenin
Monoclonal
Conc. Not Determined
ascites with 0.1% sodium azide
O60716, P30999
CTNND1
Recombinant fusion protein derivedfrom the N-terminus of murine p120ctn
RUO
12388, 1500, 311163, 428852, 483489
-20°C
Liquid
ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
6H11
Unconjugated
CTNND1
AA409437; AU019353; cadherin-associated Src substrate; Cadherin-associated Src substrate (CAS); CAS; Catenin (cadherin associated protein) delta 1; catenin (cadherin associated protein), delta 1; catenin (cadherin-associated protein), delta 1; catenin delta 1; catenin delta-1; catenin src; catenin, delta 1; Catns; CTNND; Ctnnd1; hypothetical protein; Kiaa0384; mKIAA0384; p120; p120 catenin; P120 CTN; P120 p120; P120 P120CTN; p120(CAS); p120(ctn); P120CAS; p120-catenin; P120CTN
Mouse
100 μL
Primary
Canine, Chicken, Human, Mouse, Rat
Antibody
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers