Learn More
MTCP1 Monoclonal Antibody (1G12), Invitrogen™
Mouse Monoclonal Antibody
Marca:  Invitrogen MA519372
Detalles adicionales : Peso : 0.00010kg
Código de producto. 15598802
  • 470.00€ / 100 microgramos
Fecha estimada de envío: 23-04-2024
para ver el stock
Descripción
Descripción
Peptide Sequence: MPQKDPCQKQ ACEIQKCLQA NSYMESKCQA VIQELRKCCA QYPKGRSVVC SGFEKEEEEN LTRKSASK
MTCP1 Monoclonal antibody specifically detects MTCP1 in Human samples. It is validated for ELISA, Western Blot.
Especificaciones
Especificaciones
MTCP1
Monoclonal
1 mg/mL
PBS with no preservative; pH 7.4
P56278
MTCP1
MTCP1 (AAH02600, 1 a.a. ∽ 68 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Primary
Human
Antibody
IgG2b κ
ELISA, Western Blot
1G12
Unconjugated
MTCP1
C6.1b; C61B; Cmc4; mature T cell proliferation 1; mature T-cell proliferation 1; mature T-cell proliferation 1 isoform p13; mature T-cell proliferation 1 neighbor protein; mature T-cell proliferation-1 type A; mature T-cell proliferation-1 type B1; MTCP1; MTCP-1 type A; MTCP-1 type B1; p13MTCP1; p8MTCP1; Protein p13 MTCP-1; protein p8 MTCP-1
Mouse
Affinity chromatography
RUO
4515
-20° C, Avoid Freeze/Thaw Cycles