Learn More
MTA3 Rabbit anti-Bovine, Canine, Equine, Human, Mouse, Rabbit, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Marca:  Invitrogen PA540670
Detalles adicionales : Peso : 0.00010kg
Código de producto. 15544322
  • 363.00€ / 100 microlitros
Fecha estimada de envío
23-07-2020
Añadir a la cesta
Descripción
Descripción
Plays a role in maintece of the normal epithelial architecture through the repression of SNAI1 transcription in a histone deacetylase-dependent manner, and thus the regulation of E-cadherin levels.
Especificaciones
Especificaciones
MTA3
Polyclonal
Unconjugated
MTA3
Liquid
MTA3
Synthetic peptide directed towards the N-terminal of human MTA3
Affinity Chromatography
RUO
Antibody
Polyclonal
Bovine, Canine, Equine, Human, Mouse, Rabbit, Rat
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
A6QL72, Q9BTC8, Q924K8
metastasis associated family, member 3, metastasis associated gene family, member 3, metastasis-associated protein MTA3
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
100069316, 100362346, 103347693, 116871, 475740, 512854, 57504