missing translation for 'onlineSavingsMsg'
Learn More

MSH6, clone: 44, Invitrogen™

Mouse Monoclonal Antibody

Marca:  Invitrogen 081374

Detalles adicionales : Peso : 0.04000kg

Código de producto. 10586823

  • 524.00€ / cada uno


Stock EU 2
Añadir a la cesta

Especificaciones

Especificaciones

MSH6
Monoclonal
Unconjugated
MSH6
Liquid
MSH6
Fusion protein derived from human muscle.
Purified
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin)
44
PBS with 1% BSA and 0.1% sodium azide; pH 7.4
P52701
DNA mismatch repair protein Msh6, hMSH6, G/T mismatch-binding protein, GTBP, GTMBP, MutS-alpha 160kDa subunit, p160, MSH6, GTBP
Murine
IgG1
6mL
Store at 4° C
Primary
2956
Videos
Documentos