missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MS4A7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA545837
Detalles adicionales : Peso : 0.00010kg
Descripción
Peptide sequence: TGSLSIISGK QSTKPFDLSS LTSNAVSSVT AGAGLFLLAD SMVALRTASQ Sequence homology: Dog: 79%; Human: 100%
MS4A7 Polyclonal antibody specifically detects MS4A7 in Human samples. It is validated for Western Blot.Especificaciones
MS4A7 | |
Polyclonal | |
Unconjugated | |
MS4A7 | |
4SPAN2; CD20 antigen-like 4; CD20/Fc-epsilon-RI-beta family member 4; CD20L4; CFFM4; Four-span transmembrane protein 2; high affinity immunoglobulin epsilon receptor beta subunit; membrane spanning 4-domains A7; membrane-spanning 4-domains subfamily A member 7; membrane-spanning 4-domains subfamily A member 7; LOW QUALITY PROTEIN: membrane-spanning 4-domains subfamily A member 7; membrane-spanning 4-domains, subfamily A, member 7; MGC22368; MS4A7; MS4A8 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
58475 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9GZW8 | |
MS4A7 | |
Synthetic peptide directed towards the middle region of human MS4A7 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |