missing translation for 'onlineSavingsMsg'
Learn More

S100A9 Mouse anti-Human, Clone: 1H9, Invitrogen™

Mouse Monoclonal Antibody

Marca:  Invitrogen MA140222

Detalles adicionales : Peso : 0.01000kg

Código de producto. 11569722

  • 574.00€ / 100 microgramos


Fecha de envío estimada
del proveedor 06-11-2019
Añadir a la cesta

Descripción

Descripción

MRP-14 is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein may function in the inhibition of casein kinase and altered expression of this protein is associated with the disease cystic fibrosis.
Especificaciones

Especificaciones

S100A9
Monoclonal
0.1 mg/mL
PBS with 0.1% BSA and 0.02% sodium azide
P06702
S100A9
Mouse
IgG1
100 μg
4° C
Primary
6280
ELISA, Flow Cytometry, Western Blot
1H9
Unconjugated
S100A9
Liquid
S100A9
KPpL epitope on Human MRP-14.
Purified
RUO
Antibody
Monoclonal
Human
Videos
Documentos