missing translation for 'onlineSavingsMsg'
Learn More

MORF4L2 Goat anti-Canine, Human, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Marca:  Invitrogen PA131240

Detalles adicionales : Peso : 0.01000kg

Código de producto. 11546023

  • 410.00€ / 50 microgramos


Fecha de envío estimada
del proveedor 19-06-2020
Añadir a la cesta

Especificaciones

Especificaciones

MORF4L2
Polyclonal
Unconjugated
MORF4L2
Liquid
MORF4L2
synthetic peptide SSRKQGSQPRGQQS (Human) (N terminal).
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Canine, Human
ELISA, Western Blot
0.5 mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Q15014
RP5-1055C14.2, KIAA0026, MORFL2, MRGX
Goat
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
481001, 9643
Videos
Documentos