missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MPG Monoclonal Antibody (2C5), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA519353
Detalles adicionales : Peso : 0.00010kg
Descripción
Peptide Sequence: MPARSGAQFC RRMGQKKQRP ARAGQPHSSS DAAQAPAEQP HSSSDAAQAP CPRERCLGPP TTPGPYRSIY FSSPKGHLTR LGLEFFDQPA
3′ end of the Mid1 gene, localized 68 kb upstream the humanzeta globin gene on 16p,CRA36.1 (3-methyl-adenine DNA glycosylase),N-methylpurine-DNA glycosylase, MPG,alkyladenine DNA glycosylase,proliferation-inducing protein 11,proliferation-inducing proteinEspecificaciones
MPG | |
Monoclonal | |
1.0 mg/mL | |
PBS with no preservative; pH 7.4 | |
P29372 | |
MPG | |
MPG (AAH14991, 1 a.a. ∽ 90 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
2C5 | |
Unconjugated | |
MPG | |
MDG; N-methylpurine-DNA glycosylase; AAG; APNG; CRA36.1; Mid1; N-methylpurine-DNA glycosylase, MPG; PIG11; PIG16; anpg; 3′ end of the Mid1 gene, localized 68 kb upstream the humanzeta globin gene on 16p; CRA36.1 (3-methyl-adenine DNA glycosylase); alkyladenine DNA glycosylase; proliferation-inducing protein 11; proliferation-inducing protein 16 | |
Mouse | |
Affinity chromatography | |
RUO | |
4350 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |