missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MOSPD1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA534529
Detalles adicionales : Peso : 0.01000kg
Descripción
A suggested positive control is human brain tissue lysate. PA5-34529 can be used with blocking peptide PEP-1572.
MOSPD1 Polyclonal antibody specifically detects MOSPD1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blot.Especificaciones
MOSPD1 | |
Polyclonal | |
Unconjugated | |
MOSPD1 | |
1810018L05Rik; DJ473B4; Mospd1; motile sperm domain containing 1; motile sperm domain-containing protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
317312, 56180, 70380 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q5RJS6, Q8VEL0, Q9UJG1 | |
MOSPD1 | |
A 19 amino acid peptide near the center of human MOSPD1 | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |