missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD146 Monoclonal Antibody (OJ79), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen 357800
Detalles adicionales : Peso : 0.50000kg
Descripción
CD146 Monoclonal antibody specifically detects CD146 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Western Blot.Especificaciones
CD146 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.1% sodium azide; pH 7.4 | |
P43121, Q8R2Y2, Q9EPF2 | |
MCAM | |
Recombinant human MUC18 (D1-D5) Fc protein | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Flow Cytometry, Immunohistochemistry, Western Blot | |
OJ79 | |
Unconjugated | |
MCAM | |
1-gicerin; AV025631; CD146; CD149; Cell surface glycoprotein MUC18; cell surface glycoprotein P1H12; Gicerin; HEMCAM; l-gicerin; l-gicerin precursor; l-gicerin protein; LOW QUALITY PROTEIN: cell surface glycoprotein MUC18; MCAM; melanoma adhesion molecule; melanoma cell adhesion molecule; Melanoma-associated antigen A32; melanoma-associated antigen MUC18; Muc18; mucin-18; s-endo; S-endo 1 endothelial-associated antigen; s-gicerin; s-gicerin precursor | |
Mouse | |
Protein A | |
RUO | |
4162, 78967, 84004 | |
-20°C | |
Liquid |