missing translation for 'onlineSavingsMsg'
Learn More

KIAA0090 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marca:  Invitrogen PA531597

Detalles adicionales : Peso : 0.01000kg

Código de producto. 13416127

  • 514.00€ / 100 microlitros
Fecha estimada de envío: 23-04-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Recommended positive controls: A549, HeLa, HepG2, HCT116. Predicted reactivity: Mouse (94%), Rat (94%), Rhesus Monkey (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

KIAA0090 Polyclonal antibody specifically detects KIAA0090 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.
Especificaciones

Especificaciones

KIAA0090
Polyclonal
Unconjugated
EMC1
2700016F22Rik; CAVIPMR; Emc1; ER membrane protein complex subunit 1; KIAA0090; novel DUF1620 domain containing protein; PSEC0263; RGD1310427; RP1-43E13.1
Rabbit
Antigen affinity chromatography
RUO
23065
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Q8N766
EMC1
Recombinant fragment corresponding to a region within amino acids 12 and 289 of Human KIAA0090
100 μL
Primary
Human
Antibody
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales