missing translation for 'onlineSavingsMsg'
Learn More

Ki-67 Mouse, PE-Cy7, Clone: B56, BD

Mouse Monoclonal Antibody

Marca:  BD Biosciences 561283

Detalles adicionales : Peso : 0.01000kg

Código de producto. 15810919

  • 449.00€ / cada uno
Fecha estimada de envío: 30-04-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

The B56 monoclonal antibody specifically binds to the Ki-67 antigen that is expressed in the nucleus of cycling cells (G1, S, G2, M cell cycle phases). During the G0 phase, the antigen cannot be detected. During interphase of the cell cycle, it is associated with nucleolar components, and it is on the surface of the chromosomes during M phase. Ki-67 is a large protein having 2 alternatively spliced isoforms, an N-terminal forkhead-associated domain, a C-terminal domain that binds to heterochromatin proteins, and multiple phosphorylation sites, the functions of which are still unclear. Because of the strict association of Ki-67 expression with cell proliferation, anti-Ki-67 antibodies are useful for the identification, quantification, and monitoring of growing cell populations.

Especificaciones

Especificaciones

Ki-67
Monoclonal
PE-Cyanine7
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Human Ki-67
50 Tests
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Flow Cytometry
B56
MKI67; Antigen identified by monoclonal antibody Ki-67; KIA
Mouse
Affinity Purified
RUO
Human, Mouse, Rat, Rhesus Monkey
IgG1 κ
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales