missing translation for 'onlineSavingsMsg'
Learn More

KCNT2 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marca:  Invitrogen PA523855

Detalles adicionales : Peso : 0.01000kg

Código de producto. 13208408

  • 514.00€ / 100 microgramos
Fecha estimada de envío: 23-04-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

KCNT2 Polyclonal antibody specifically detects KCNT2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.
Especificaciones

Especificaciones

KCNT2
Polyclonal
Unconjugated
Kcnt2
E330038N15Rik; KCa4.2; Kcnt2; Potassium channel subfamily T member 2; potassium channel, sodium activated subfamily T, member 2; potassium channel, sodium-activated subfamily T, member 2; potassium channel, subfamily T, member 2; potassium sodium-activated channel subfamily T member 2; sequence like an intermediate conductance potassium channel subunit; SLICK; SLO2.1; sodium- and chloride-activated ATP-sensitive potassium channel; sodium and chloride-activated ATP-sensitive potassium channel Slo2.1; sodium- and chloride-activated ATP-sensitive potassium channel subunit; sodium-and chloride-activated ATP-sensitive potassium channel (SLICK)
Rabbit
Antigen affinity chromatography, Protein A
RUO
343450
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
0.4 mg/mL
PBS with 0.09% sodium azide
Q6UVM3
Kcnt2
KLH conjugated synthetic peptide between 1069-1097 amino acids from the C-terminal region of human KCNT2
400 μL
Primary
Human
Antibody
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales