missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KANK3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA534540
Detalles adicionales : Peso : 0.01000kg
Descripción
A suggested positive control is A431 cell lysate. PA5-34540 can be used with blocking peptide PEP-1583.
KANK3 Polyclonal antibody specifically detects KANK3 in Human, Mouse samples. It is validated for Immunohistochemistry, Western Blot.Especificaciones
KANK3 | |
Polyclonal | |
Unconjugated | |
KANK3 | |
0610013D04Rik; Ankrd47; ankyrin repeat domain 47; ankyrin repeat domain-containing protein 47; D17Ertd288e; Kank3; kidney ankyrin repeat-containing protein 3; KN motif and ankyrin repeat domain-containing protein 3; KN motif and ankyrin repeat domains 3; NG28; putative KN motif and ankyrin repeat domains 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
256949, 80880 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q6NY19, Q9Z1P7 | |
KANK3 | |
An 18 amino acid peptide near the amino terminus of human KANK3 | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |