Learn More
ITM2B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA531441
Detalles adicionales : Peso : 0.01000kg
Descripción
Recommended positive controls: HepG2, HCT116, mouse brain, rat brain. Predicted reactivity: Mouse (95%), Rat (96%), Pig (95%), Sheep (96%), Rhesus Monkey (100%), Chimpanzee (100%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ITM2B Polyclonal antibody specifically detects ITM2B in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Especificaciones
ITM2B | |
Polyclonal | |
Unconjugated | |
ITM2B | |
ABRI; ABri/ADan amyloid peptide; ABri23; AI256040; Bri; BRI2; BRI2 ICD; BRI2 intracellular domain; BRI2, membrane form; Bri2-23; Bri23 peptide; BRI2C, soluble form; Bricd2b; BRICHOS domain containing 2B; C-terminal peptide; D14Sel6; E25B; E25BMM; E3-16; FBD; imBRI2; Immature BRI2; integral membrane protein 2B; Integral membrane protein 2B (E25B protein); ITM2B; Mature BRI2; mBRI2; OTTHUMP00000018394; P23 peptide; Protein E25B; RDGCA; RP11-196P14.1; Transmembrane protein BRI | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
16432, 290364, 9445 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.81 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
O89051, Q5XIE8, Q9Y287 | |
ITM2B | |
Recombinant fragment corresponding to a region within amino acids 15 and 264 of Human ITM2B | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |