missing translation for 'onlineSavingsMsg'
Learn More

ITGB1BP2 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marca:  Invitrogen PA542154

Detalles adicionales : Peso : 0.00010kg

Código de producto. 15505292

  • 473.00€ / 100 microlitros
Fecha estimada de envío: 08-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Peptide sequence: MSLLCRNKGC GQHFDPNTNL PDSCCHHPGV PIFHDALKGW SCCRKRTVDF Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 86%; Horse: 93%; Human: 100%; Mouse: 100%; Pig: 93%; Rabbit: 100%; Rat: 100%

ITGB1BP2 Polyclonal antibody specifically detects ITGB1BP2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.
Especificaciones

Especificaciones

ITGB1BP2
Polyclonal
Unconjugated
ITGB1BP2
Chordc3; integrin beta 1 binding protein (melusin) 2; integrin beta 1 binding protein 2; integrin beta-1-binding protein 2; integrin subunit beta 1 binding protein 2; ITGB1BP; Itgb1bp2; Melusin; MGC119214; MSTP015; RGD1565015
Rabbit
Protein A
RUO
26548
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q9UKP3
ITGB1BP2
Synthetic peptide directed towards the N-terminal of human ITGB1BP2
100 μL
Primary
Human
Antibody
IgG
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali