missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD103 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA523996
Detalles adicionales : Peso : 0.01000kg
Descripción
CD103 Polyclonal antibody specifically detects CD103 in Human samples. It is validated for Western Blot.Especificaciones
CD103 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
P38570 | |
Itgae | |
KLH conjugated synthetic peptide between 1146-1173 amino acids from the C-terminal region of human ITGAE | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
Itgae | |
A530055J10; alpha-E1; alpha-M290; aM290; antigen CD103; antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide; CD103; DC; DCs; HML-1 antigen; human mucosal lymphocyte antigen 1, alpha polypeptide; HUMINAE; integrin alpha E; integrin alpha E, epithelial-associated; integrin alpha E1, epithelial-associated; integrin alpha M290; Integrin alpha-E; Integrin alpha-E heavy chain; Integrin alpha-E light chain; Integrin alpha-IEL; integrin subunit alpha E; integrin, alpha E; integrin, alpha E (antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide); integrin, alpha E, epithelial-associated; Intregrin alpha E2; Itgae; MGC141996; Mucosal lymphocyte 1 antigen | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
3682 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |