Learn More
ITGA4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA532882
Detalles adicionales : Peso : 0.01000kg
Descripción
In IHC applications, antigen retrieval using a citrate buffer (pH 6.0) is recommended. Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Mouse, Rat, Dog, Bat, Hamster, Panda, Rabbit, Pig (100%).
ITGA4 Polyclonal antibody specifically detects ITGA4 in Hamster, Human, Mouse, Non-human Primate, Porcine, Rabbit, Rat samples. It is validated for Immunohistochemistry (Paraffin).Especificaciones
ITGA4 | |
Polyclonal | |
Unconjugated | |
ITGA4 | |
269C wild type; alpha 4 subunit of VLA-4 receptor; antigen CD49D, alpha-4 subunit of VLA-4 receptor; CD49 antigen-like family member D; CD49d; IA4; integrin alpha 4; integrin alpha-4; integrin alpha4B; Integrin alpha-IV; integrin subunit alpha 4; integrin, alpha 4; integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor); Itga4; Itga4B; ITGA-IV; LPAM subunit alpha; lymphocyte Peyer patch adhesion molecules subunit alpha; very late activation protein 4 receptor, alpha 4 subunit; VLA-4 receptor, alpha 4 subunit; VLA-4 subunit alpha | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100346103, 100521477, 100762323, 16401, 311144, 3676 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P13612, Q00651 | |
ITGA4 | |
Synthetic 18 amino acid peptide from C-terminus of human ITGA4 / Integrin Alpha 4. | |
50 μg | |
Primary | |
Hamster, Human, Mouse, Non-human Primate, Porcine, Rabbit, Rat | |
Antibody | |
IgG |