Learn More
IRAK4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA529699
Detalles adicionales : Peso : 0.01000kg
Descripción
Recommended positive controls: H1299. Predicted reactivity: Mouse (89%), Rat (90%), Pig (95%), Sheep (93%), Rhesus Monkey (100%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
IRAK4 Polyclonal antibody specifically detects IRAK4 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Especificaciones
IRAK4 | |
Polyclonal | |
Unconjugated | |
IRAK4 | |
8430405M07Rik; 9330209D03Rik; interleukin 1 receptor associated kinase 4; interleukin-1; interleukin-1 receptor-associated kinase 4; IPD1; Irak4; IRAK-4; NY-REN-64; REN64; renal carcinoma antigen NY-REN-64 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51135 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.54 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q9NWZ3 | |
IRAK4 | |
Recombinant fragment corresponding to a region within amino acids 215 and 456 of Human IRAK4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |