Learn More
IP3 Receptor 1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA3901A
Detalles adicionales : Peso : 0.01000kg
Description
PA3-901A detects inositol 1,4,5-triphosphate receptor type-I (IP3R-I) from mouse, human and rat tissues. PA3-901A has been successfully used in Western blot procedures. By Western blot, this antibody detects a ∽240-260 kDa protein representing IP3R-I from rat and mouse brain extract. PA3-901A is a remanufactured lot of PA3-901 which has previously been used in Western blot, immunoprecipitation and immunohistochemistry procedures. The PA3-901A immunogen is a synthetic peptide corresponding to residues N(1829) K K K D D E V D R D A P S R K K A K E(1848) of human IP3 Receptor Type I. This sequence is 100% conserved in human, mouse, and rat IP3R-I protein. This peptide (Cat. # PEP-019) is available for use in neutralization and control experiments.
IP3 Receptor 1 Polyclonal antibody specifically detects IP3 Receptor 1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
IP3 Receptor 1 | |
Polyclonal | |
Unconjugated | |
Itpr1 | |
ACV; CLA4; D6Pas2; ENSMUSG00000072853; Gm10429; I145TR; inositol 1,4,5-triphosphate receptor 1; inositol 1,4,5-triphosphate receptor, type 1; inositol 1,4,5-trisphosphate receptor 1; inositol 1,4,5-trisphosphate receptor type 1; inositol 1,4,5-trisphosphate receptor, type 1; inositol 1,4,5-trisphosphate-binding protein P400; InsP>3<R type I; Insp3r; InsP3R type I; insP3R1; IP3 receptor; IP3 receptor isoform 1; Ip3r; IP-3-R; IP3R 1; IP3R1; Itpr1; Itpr-1; opisthotonus; opt; P400; Pcd6; Pcp1; Pcp-1; PPP1R94; protein PCD-6; protein phosphatase 1, regulatory subunit 94; Purkinje cell protein 1; SCA15; SCA16; SCA29; SI-SIII-SIIA; Type 1 inositol 1,4,5-trisphosphate receptor; type 1 InsP3 receptor | |
Rabbit | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Western Blot | |
Conc. Not Determined | |
whole serum diluted in PBS with 0.05% sodium azide | |
P11881, P29994, Q14643 | |
Itpr1 | |
Synthetic peptide corresponding to residues N(1829) K K K D D E V D R D A P S R K K A K E(1848) of human IP3 Receptor Type I. | |
RUO | |
16438, 25262, 3708 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |