Learn More
IL-8 (CXCL8) Polyclonal Antibody, Biotin, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA186581
Detalles adicionales : Peso : 0.01000kg
Descripción
Reconstitute with 0.5 mL PBS containing 0.1% BSA. For long term storage the addition of 0.09% sodium azide is recommended. Prior to reconstitution, store at 4oC. After reconstitution, store undiluted at -20oC, avoiding freeze/thaw cycles. This biotinylated human IL-8 antibody may be used in a direct ELISA or as the detection reagent in a sandwich ELISA with PA1-86581 as the capture reagent. Rabbit anti Human Interleukin-8 antibody recognizes human IL-8 (Interleukin-8), a 72 amino acid pro-inflammatory CXC chemokine designated CXCL8, secreted by endothelial cells and monocytes, which acts as a major chemoattractant for neutrophils and also T cells and basophils.
IL-8 (CXCL8) Polyclonal antibody specifically detects IL-8 (CXCL8) in Human samples. It is validated for ELISA, Western Blot.Especificaciones
IL-8 (CXCL8) | |
Polyclonal | |
Biotin | |
CXCL8 | |
(Ala-IL-8)77; (Ser-IL-8)72; Alveolar macrophage chemotactic factor I; alveolar macrophage-derived chemotactic factor-I; AMCF-I; ANX7; beta endothelial cell-derived neutrophil activating peptide; beta-thromboglobulin-like protein; chemokine (C-X-C motif) ligand 8; C-X-C motif chemokine 8; C-X-C motif chemokine ligand 8; CXCL8; cytokine 8; emoctakin; GCP/IL-8 protein I; GCP/IL-8 protein II; GCP/IL-8 protein III; GCP/IL-8 protein IV; GCP/IL-8 protein V; GCP/IL-8 protein VI; GCP1; GCP-1; Granulocyte chemotactic protein 1; IL8; IL-8; IL-8(1-77); IL-8(5-77); IL-8(6-77); IL-8(7-77); IL-8(8-77); IL-8(9-77); IL8/NAP1 form I; IL8/NAP1 form II; IL8/NAP1 form III; IL8/NAP1 form IV; IL8/NAP1 form V; IL8/NAP1 form VI; ILN; Interleukin; interleukin 8; Interleukin8; interleukin-8; interleuklin-8 precursor; LECT; LUCT; lung giant cell carcinoma-derived chemotactic protein; lymphocyte derived neutrophil activating peptide; Lymphocyte-derived neutrophil-activating factor; LYNAP; MDNCF; MDNCF-a; MDNCF-b; | |
Rabbit | |
Affinity chromatography | |
RUO | |
3576 | |
-20°C | |
Lyophilized |
ELISA, Western Blot | |
0.1 mg/mL | |
PBS with no preservative | |
P10145 | |
CXCL8 | |
Recombinant human IL-8 | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |