Learn More
IL-4 Mouse anti-Human, PE, Clone: 8D4-8, BD
Mouse Monoclonal Antibody
Marca: BD Biosciences 562046
Detalles adicionales : Peso : 0.02000kg
Descripción
The 8D4-8 monoclonal antibody reacts with human interleukin-4 (IL-4). The immunogen used to raise the 8D4-8 hybridoma was recombinant human IL-4. The 8D4-8 antibody binds to an epitope that is different than the epitope recognized by the MP4-25D2 antibody (Cat. No. 554485).
Clone 8D4-8 displays an increased amount of non-specific binding to dead cells when compared to the clone MP4-25D2. It is recommended to use a fixable viability dye in conjunction with this clone.
Especificaciones
IL-4 | |
Monoclonal | |
PE | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |
Flow Cytometry | |
8D4-8 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Recombinant Human IL-4 | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.