Learn More
IL-4 Rat anti-Human, Brilliant Violet 421, Clone: MP4-25D2, BD
Rat Monoclonal Antibody
Marca: BD Biosciences 564110
Detalles adicionales : Peso : 0.02000kg
Descripción
The MP4-25D2 antibody reacts with human interleukin-4 (IL-4). The immunogen used to generate the MP4-25D2 hybridoma was purified recombinant human IL-4. This is a neutralizing antibody. The MP4-25D2 antibody has been reported to cross react with IL-4 from rhesus monkeys. The use of the MP4-25D2 antibody for epitope mapping of human IL-4 has been described.
The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon™ Brilliant Violet™ family of dyes. With an Ex Max of 407-nm and Em Max at 421-nm, BD Horizon™ BV421can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50-nm filter). BD Horizon™ BV421conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue™ conjugates.
Especificaciones
IL-4 | |
Monoclonal | |
Brilliant Violet 421 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Purified Recombinant Human IL-4 | |
100 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
MP4-25D2 | |
Interleukin-4; IL4; BCGF-1; BSF-1; Lymphocyte stimulatory factor 1 | |
Rat | |
Affinity Purified | |
RUO | |
Human | |
IgG1 |