missing translation for 'onlineSavingsMsg'
Learn More

anti-IgG3 κ, Mouse, Clone: J606, Isotype Control, BD

Marca:  BD Biosciences 553466

Detalles adicionales : Peso : 0.01000kg

Código de producto. 15827238

  • 680.00€ / cada uno
Fecha estimada de envío: 18-04-2024
para ver el stock
Añadir a la cesta

Descripción

Descripción

Mouse IgG3 κ isotype control antibody is specific for the β-2 - 6-linked Fructosan. The transplantable plasmacytoma J606 was induced by inraperitoneal injection of mineral oils into BALB/c mice. This antibody is routinely tested by immunohistochemistrical analysis on frozen rat tissue sections. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Mouse
Clone: J606
Isotype: IgG3, κ

Immunohistochemistry (Formalin-Antigen Retrieval Required), Immunohistochemistry (Frozen), Immunohistochemistry (Zinc-Fixed), Isotype Control

Especificaciones

Especificaciones

Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Zinc-Fixed)
1mg/mL
Mouse
0.5 mg
IgG3 κ
J606
Unconjugated
Affinity Purified
Undiluted at 4°C
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales