missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Human IgE Secondary Antibody, PE, Invitrogen™
Mouse Monoclonal Secondary Antibody
Marca: Invitrogen MA110375
Detalles adicionales : Peso : 0.01000kg
Descripción
This antibody recognizes an epitope different from the ones recognized by clones 4G7 and 4H10.
Human IgE Monoclonal antibody specifically detects Human IgE in Human samples. It is validated for Flow Cytometry.Especificaciones
Human IgE | |
Monoclonal | |
0.1 mg/mL | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
Purified human IgE. | |
100 μg | |
Secondary | |
4° C, store in dark, DO NOT FREEZE! | |
IgG1 |
Flow Cytometry | |
BE5 | |
PE | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
Human | |
Liquid |