missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IkB alpha Monoclonal Antibody (ZI002), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen 397700
Detalles adicionales : Peso : 0.50000kg
Descripción
IkB alpha Monoclonal antibody specifically detects IkB alpha in Human, Mouse samples. It is validated for ELISA, Immunocytochemistry, Western Blot.Especificaciones
IkB alpha | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.1% sodium azide; pH 7.4 | |
P25963, Q9Z1E3 | |
NFKBIA | |
Synthetic peptide derived from the C-terminal region of the human I-kappa-B-alpha protein | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 κ |
ELISA, Immunocytochemistry, Western Blot | |
ZI002 | |
Unconjugated | |
NFKBIA | |
AI462015; ECI-6; ECI-6/IKBA; I kappa B-alpha; I(Kappa)B(alpha); I79_018146; IkappaB alpha; IkappaBalpha; I-kappaBalpha; I-kappa-B-alpha; Ikba; IKBalpha; IKB-alpha; Inhibitor of nuclear factor of kappa light chain gene enhancer in B-cells alpha; Inhibitor of nuclear factor of kappa light chain gene enhancer in B-cells, alpha; MAD3; MAD-3; Major histocompatibility complex enhancer-binding protein MAD3; NF kappa B inhibitor alpha; NF-kappaB inhibitor alpha; NF-kappa-B inhibitor alpha; NFKB inhibitor alpha; NFKBI; Nfkbia; nuclear factor of kappa light chain gene enhancer in B-cells; nuclear factor of kappa light chain gene enhancer in B-cells inhibitor, alpha; nuclear factor of kappa light polyp gene enhancer in B-cell 1; nuclear factor of kappa light polypeptide gene enhancer in B cells inhibitor, alpha; nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, alpha; Rel-associated pp40; REL-associated protein pp40; RL/IF-1 | |
Mouse | |
Protein A | |
RUO | |
18035, 4792 | |
-20°C | |
Liquid |