Learn More
HS1 Mouse anti-Human, Unlabeled, Clone: 9, BD
Mouse Monoclonal Antibody
Marca: BD Biosciences 610541
Detalles adicionales : Peso : 0.01000kg
Descripción
HS1 (lck-binding protein 1 (lckBP1)) is a protein of 486 amino acids with homology to cytoplasmic tyrosine kinases. The protein contains an SH3 domain and two proline-rich regions, that may be the binding site for lck. Co- mmunoprecipitation experiments demonstrated a tight association between HS1 and lck. The SH3 domain of lck is necessary for this interaction. In addition, HS1 contains tandem repeat sequence of seven amino acids which is a substrate for the tyrosine kinase src. This sequence is also found in the protein cortactin. Therefore, HS1, like cortactin, may be an important adaptor that is critical for the regulation of cellular tyrosine kinases.
This antibody is routinely tested by western blot analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Especificaciones
HS1 | |
Monoclonal | |
250μg/mL | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
150 μg | |
Cell Biology | |
Human | |
IgG1 |
Western Blot | |
9 | |
Unconjugated | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.