missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXD10 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marca: Invitrogen PA519007
Detalles adicionales : Peso : 0.01000kg
Descripción
This antibody is predicted to react with bovine, equine, mouse and rat based on sequence homology.
HOXD10 Polyclonal antibody specifically detects HOXD10 in Human samples. It is validated for Immunohistochemistry, Western Blot.Especificaciones
HOXD10 | |
Polyclonal | |
Unconjugated | |
HOXD10 | |
AI385591; AI874987; d10; homeo box 4D; homeo box D10; homeo box D10a; homeobox D10; homeobox D10a; homeobox gene D-10; homeobox protein Hox-4.5; Homeobox protein Hox-4D; homeobox protein Hox-4E; Homeobox protein Hox-5.3; homeobox protein Hox-C10; Homeobox protein Hox-D10; homeobox protein Hox-D10a; homeobox protein Hox-D11; homeobox protein Hox-D10; HOX4; Hox-4.4; Hox-4.5; Hox-45; HOX4D; HOX4E; Hox-5.3; hoxc10; hox-C10; Hoxc10 protein; Hoxd10; HOX-D10; hoxd-10; hoxd10a; hoxd10a protein; z-82 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
3236 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P28358 | |
HOXD10 | |
Synthetic peptide sequence (PNRSCRIEQPVTQQ) corresponding to the internal amino acids of HOXD10 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |