missing translation for 'onlineSavingsMsg'
Learn More

HMGCR Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marca:  Invitrogen PA537367

Detalles adicionales : Peso : 0.00010kg

Código de producto. 15513611

  • 550.00€ / 100 microlitros
Fecha estimada de envío: 18-04-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This antibody detects endogenous protein at a molecular weight of 97 kDa. Purity is >95% by SDS-PAGE.

HMGCR Polyclonal antibody specifically detects HMGCR in Human, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.
Especificaciones

Especificaciones

HMGCR
Polyclonal
Unconjugated
HMGCR
3H3M; 3-hydroxy-3-methylglutaryl CoA reductase (NADPH); 3-hydroxy-3-methylglutaryl-CoA reductase; 3-hydroxy-3-methylglutaryl-Coenzyme A reductase; HMDH; HMG CoA reductase; HMG CoAR; HMG-CoA reductase; HMG-CoAR; HMGCR; hydroxymethylglutaryl-CoA reductase; LDLCQ3; MGC103269; Red
Rabbit
Antigen affinity chromatography
RUO
25675, 3156
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide
P04035, P51639
HMGCR
Recombinant full length human HMGCR
100 μL
Primary
Human, Rat
Antibody
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales