missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMGB1 Polyclonal Antibody, Biotin, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA522719
Detalles adicionales : Peso : 0.01000kg
Descripción
HMGB1 Polyclonal antibody specifically detects HMGB1 in Bovine, Canine, Human, Mouse, Ovine, Rat samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Especificaciones
HMGB1 | |
Polyclonal | |
Biotin | |
Hmgb1 | |
Ac2-008; amphoterin; Amphoterin antibody; DEF; DKFZp686A04236; Heparin-binding protein p30; high mobility group 1; high mobility group 1 protein; high mobility group box 1; high mobility group protein 1; High mobility group protein B1; high mobility group protein HMG1; high-mobility group (nonhistone chromosomal) protein 1; high-mobility group box 1; high-mobility-group protein; Hmg1; HMG-1; HMG3; HMG3 antibody; hmgb 1; HMGB1; hmgb-1; HMGB1 protein; hypothetical protein; non-histone protein HMG1; p30; RCJMB04_15a21; RP11-550P23.1; SBP-1; Sulfoglucuronyl carbohydrate binding protein; unnamed protein product | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
101112071, 15289, 25459, 282691, 3146, 403170 | |
4° C, store in dark | |
Liquid |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1.10 mg/mL | |
PBS with 0.05% sodium azide | |
P09429, P10103, P63158, P63159, Q6YKA4 | |
Hmgb1, LOC101112071 | |
A synthetic peptide made to an internal portion of the human HMGB1 protein sequence (between residues 100-200). | |
100 μL | |
Primary | |
Bovine, Canine, Human, Mouse, Ovine, Rat | |
Antibody | |
IgG |