Learn More
HLA-DQ Mouse anti-Human, Unlabeled, Clone: TÜ169, BD
Mouse Monoclonal Antibody
Marca: BD Biosciences 555562
Detalles adicionales : Peso : 0.01000kg
Descripción
Reacts with major histocompatibility Class II HLA-DQ1 and DQ2 antigens and weakly with HLA-DQ3. HLA-DQ is expressed primarily on immunocompetent B cells and also on macrophages. This antibody does not cross-react with HLA-DR2 and DR7. This antibody fixes complement. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Flow Cytometry
Especificaciones
HLA-DQ | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
Flow Cytometry | |
TÜ169 | |
Unconjugated | |
Mouse | |
0.1 mg | |
Immunology | |
Human | |
IgG2a κ |
For Research Use Only.