missing translation for 'onlineSavingsMsg'
Learn More

HLA-B7 Monoclonal Antibody (BB7.1), PE, Invitrogen™

Mouse Monoclonal Antibody

Marca:  Invitrogen MA516585

333.35 EUR hasta el 2024-03-29
Use el código promocional "21615" para aplicar la oferta.



Detalles adicionales : Peso : 0.01000kg

Código de producto. 15110835

  • 500.00€ / 100 tests
Fecha estimada de envío: 23-04-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Reconstitute with 1 mL of distilled water. After reconstitution, store product undiluted at 4oC in the dark. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.

HLA-B7 Monoclonal antibody specifically detects HLA-B7 in Cynomolgus Monkey, Human samples. It is validated for Flow Cytometry.
Especificaciones

Especificaciones

HLA-B7
Monoclonal
Conc. Not Determined
PBS with 1% BSA, 5% sucrose and 0.09% sodium azide
P01889
HLA-B
Papain solubilized HLA-B7 antigen
100 Tests
Primary
Cynomolgus Monkey, Human
Antibody
IgG1
Flow Cytometry
BB7.1
PE
HLA-B
A29 protein; antigen presenting molecule; AS; Aw-33; Aw-74; B-4901; B-5001; Bw-47; Bw-50; CDABP0067; class I MHC, HLA-B antigen; DAQB-90C11.16; epididymis secretory protein Li 83; FLJ26655; HEL-S-83; histocompatibility antigen; HLA A; HLA -B; HLA B-40011; HLA B44; hla b7; HLA class I antigen; HLA class I antigen HLA-B; HLA class I histocompatibility antigen, A alpha chain; HLA class I histocompatibility antigen, A-1 alpha chain; HLA class I histocompatibility antigen, A-30 alpha chain; HLA class I histocompatibility antigen, A-33 alpha chain; HLA class I histocompatibility antigen, A-74 alpha chain; HLA class I histocompatibility antigen, B alpha chain; HLA class I histocompatibility antigen, B-47 alpha chain; HLA class I histocompatibility antigen, B-50 alpha chain; HLA class I histocompatibility antigen, B-82 alpha chain; HLA locus; HLA molecule; HLAA; HLA-A; HLA-A locus alpha 2 domain; HLA-A null; HLA-A11; HLA-A33; HLA-Aw33.1; HLAB; HLA-B; HLA-B 1502; HLA-B alpha chain; HLA-B alpha
Mouse
Protein G
RUO
3106
4° C, store in dark, DO NOT FREEZE!
Lyophilized
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales