missing translation for 'onlineSavingsMsg'
Learn More

Histone H3 Monoclonal Antibody (865R2), Invitrogen™

Mouse Monoclonal Antibody

Marca:  Invitrogen AHO1432

Detalles adicionales : Peso : 0.10000kg

Código de producto. 10399194

  • 459.00€ / cada uno
En stock
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Histone H3 Monoclonal antibody specifically detects Histone H3 in Human, Mouse, Non-human Primate samples. It is validated for Immunocytochemistry, Western Blot.
Especificaciones

Especificaciones

Histone H3
Monoclonal
0.5 mg/mL
PBS with 1% BSA and 0.1% sodium azide; pH 7.2
P68431, P68433, P84228, Q71DI3
H3C1, H3C10, H3C12, H3C14, H3C15, H3C2, H3C7, HIST1H3A
Recombinant human histone H3 expressed in E. coli.
100 μg
Primary
Human, Mouse, Non-human Primate
Antibody
IgM
Immunocytochemistry, Western Blot
865R2
Unconjugated
H3C1
BUR5; CG31613; CG31613-PA; CG33803; CG33806; CG33809; CG33812; CG33815; CG33818; CG33821; CG33824; CG33827; CG33830; CG33833; CG33836; CG33839; CG33842; CG33845; CG33848; CG33851; CG33854; CG33857; CG33860; CG33863; CG33866; CG5825; CG5825-PA; CG5825-PC; CG5825-PD; CG8989; dH3.3A; Dmel\CG31613; Dmel\CG5825; Dmel_CG31613; Dmel_CG5825; fb58e10; h3; H3 histone; H3 histone family, member A; H3 histone family, member I; H3 histone family, member K; H3 histone family, member L; H3 histone family, member M; H3 histone, family 2; H3 histone, family 3A; H3 histone, family 3B; H3 histone, family 3B (H3.3B); H3 histone, family 3B.1; H3 histone, family 3C; H3.1-221; H3.1-291; H3.1-I; H3.2; H3.2-221; H3.2-614; H3.2-615; H3.2-616; h3.2a; H3.3; H3.3 histone A; H3.3 histone B; H3.3a; H3.3B; H3.5; H3.5 histone; H3.A; H3.B; H3/A; H3/b; H3/d; H3/f; H3/i; H3/j; H3/k; H3/l; H3/M; H3/n; H3/o; H3-143; H3-291; H3-3A; H3-3B; H3-5; H3-53; H3-614; H3a; H3b; H3-B; H3C1; H3c10; H3c11; H3C12; H3C2; H3C3; H3C4; H3C6
Mouse
purified
RUO
126961, 260423, 319150, 319152, 333932, 360198, 8350, 8356, 8357, 8358, 8968, 97114
-20°C
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales