missing translation for 'onlineSavingsMsg'
Learn More

Gelsolin Mouse, Unlabeled, Clone: 2, BD

Mouse Monoclonal Antibody

Marca:  BD Biosciences 610412

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15825449

  • 241.00€ / 1 set


Fecha estimada de envío
14-11-2019
Añadir a la cesta

Descripción

Descripción

Gelsolin was identified as a result of its ability to sever actin filaments in a Ca2+-dependent manner. The gene for gelsolin encodes an 83kDa protein that migrates as a 93kDa polypeptide in SDS-gels. The N-terminal domain contains the calcium-independent actin-severin site, whereas the calcium-dependent site is located in the C-terminal portion of the protein. It exhibits significant homology with villin, another calcium-regulated actin filament severing protein. Gelsolin can be found intracellularly, as well as in a secreted form. However, both forms are encoded by the same gene.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

Especificaciones

Especificaciones

Gelsolin
2
Unconjugated
Affinity Purified
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
50μg
Cell Biology
Primary
Canine, Human, Murine, Rabbit, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG2a
RUO
Store undiluted at -20°C.
Monoclonal
Videos
Documentos

For Research Use Only.