missing translation for 'onlineSavingsMsg'
Learn More

GBA3 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marca:  Invitrogen PA530782

Detalles adicionales : Peso : 0.01000kg

Código de producto. 13455287

  • 470.00€ / 100 microlitros
Fecha estimada de envío: 23-04-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Recommended positive controls: 293T, A431, Jurkat. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

GBA3 Polyclonal antibody specifically detects GBA3 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.
Especificaciones

Especificaciones

GBA3
Polyclonal
Unconjugated
GBA3
CBG; CBGL1; Cytosolic beta-glucosidase; cytosolic beta-glucosidase-like protein 1; Cytosolic GCase; Cytosolic glycosylceramidase; Gba3; GLUC; Glucosidase beta acid 3; glucosidase, beta, acid 3; glucosidase, beta, acid 3 (cytosolic); Glucosylceramidase beta 3; glucosylceramidase beta 3 (gene/pseudogene); klotho-related protein; KLRP
Rabbit
Antigen affinity chromatography
RUO
57733
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.55 mg/mL
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Q9H227
GBA3
Recombinant protein fragment corresponding to a region within amino acids 1 and 139 of Human GBA3
100 μL
Primary
Human
Antibody
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales