missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAPDH Loading Control Monoclonal Antibody (GA1R), Alexa Fluor™ 555, Invitrogen™
Mouse Monoclonal Antibody
Marque: Invitrogen MA515738A555
Informations supplémentaires : Peso : 0.00900kg
Description
GAPDH Loading Control Monoclonal antibody specifically detects GAPDH Loading Control in Bacteria, Chicken, Hamster, Human, Insect, Mouse, Rabbit, Rat, Yeast samples. It is validated for Immunocytochemistry, Western Blot.Spécification
GAPDH Loading Control | |
Monoclonal | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide | |
O14556, P00356, P04406, P04797, P16858, P17244, P46406, Q64467, Q9ESV6 | |
GAPDH, GAPDHS | |
recombinant GAPDH | |
50 μL | |
Primary | |
Bacteria, Chicken, Hamster, Human, Insect, Mouse, Rabbit, Rat, Yeast | |
Antibody | |
IgG1 |
Immunocytochemistry, Western Blot | |
GA1R | |
Alexa Fluor 555 | |
GAPDHS | |
38 kDa BFA-dependent ADP-ribosylation substrate; aging-associated gene 9 protein; BARS-38; bb02e05; cb350; cb609; CDABP0047; EC 1.2.1.12; epididymis secretory protein Li 278; epididymis secretory sperm binding protein Li 162eP; fb71f08; fk58c09; G3PD; G3PDH; GAPD; GAPD2; gapdh; GAPDH2; GAPDH-2; GAPDHS; Gapds; Gapd-s; glceraldehyde-3-phosphate dehydrogenase; glyceraldehyde 3-phosphate dehydrogenase; glyceraldehyde 3-phosphate dehydrogenase, testis-specific; glyceraldehyde phosphate dehydrogenase; glyceraldehyde-3-phosphate dehydrogenase; glyceraldehyde-3-phosphate dehydrogenase (G3PDH); glyceraldehyde-3-phosphate dehydrogenase 2; glyceraldehyde-3-phosphate dehydrogenase GAPDH; glyceraldehyde-3-phosphate dehydrogenase like-17 protein; glyceraldehyde-3-phosphate dehydrogenase type 2; glyceraldehyde-3-phosphate dehydrogenase, spermatogenic; glyceraldehyde-3-phosphate dehydrogenase, testis-specific; glyceraldehyde-phosphate-dehydrogenase; glycerine aldehyde 3-phosphate dehydrogenase; HEL-S- | |
Mouse | |
Protein A | |
RUO | |
100009074, 100352213, 100736557, 100767862, 14433, 14447, 24383, 2597, 26330, 374193, 66020 | |
4° C, do not freeze | |
Liquid |