missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAP43 Polyclonal Antibody, DyLight™ 488, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA522897
Detalles adicionales : Peso : 0.01000kg
Descripción
Suggested positive control: embryonic nervous tissue, brain, injured nerve.
GAP43 Polyclonal antibody specifically detects GAP43 in Bovine, Canine, Chicken, Drosophila, Equine, Human, Mouse, Non-human Primate, Porcine, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.Especificaciones
GAP43 | |
Polyclonal | |
DyLight 488 | |
GAP43 | |
Axonal membrane protein GAP-43; B-50; Basp2; brain abundant, membrane attached signal protein 2; calmodulin-binding protein P-57; Gap43; GAP-43; growth accentuating protein 43; growth associated protein 43; growth-associated protein 43; MGC156666; nerve growth-related peptide GAP43; neural phosphoprotein B-50; Neuromodulin; neuron growth-associated protein 43; neuron protein F1; PP46; Protein F1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100061125, 100623423, 14432, 2596, 281777, 29423, 427955, 478572 | |
4° C, store in dark | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
0.67 mg/mL | |
50mM sodium borate with 0.05% sodium azide | |
P06836, P06837, P07936, P17677, P35001 | |
GAP43 | |
Synthetic peptide corresponding to amino acids 216-226 of rat/mouse GAP-43, (C)-KEDPEADQEHA coupled to maleimide-activated KLH. | |
100 μL | |
Primary | |
Bovine, Canine, Chicken, Drosophila, Equine, Human, Mouse, Non-human Primate, Porcine, Rat | |
Antibody | |
IgG |