Learn More
CSGALNACT2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA543328
Detalles adicionales : Peso : 0.00010kg
Descripción
Peptide sequence: LYNPAIVYAN QEVPPPVEQQ LVHKKDSGFW RDFGFGMTCQ YRSDFLTIGG Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 77%
CSGALNACT2 Polyclonal antibody specifically detects CSGALNACT2 in Human samples. It is validated for Western Blot.Especificaciones
CSGALNACT2 | |
Polyclonal | |
Unconjugated | |
Csgalnact2 | |
4632415D10Rik; beta 4 GalNAcT-2; beta4GalNAcT; Beta4GalNAcT-2; CHGN2; ChGn-2; chondroitin beta1,4 N-acetylgalactosaminyltransferase 2; Chondroitin beta-1,4-N-acetylgalactosaminyltransferase 2; chondroitin sulfate GalNAcT-2; chondroitin sulfate N-acetylgalactosaminyltransferase 2; CSGALNACT2; Galnact2; galNAcT-2; glucuronylgalactosylproteoglycan 4-beta-N-acetylgalactosaminyltransferase 2; PRO0082; RGD1563660 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
55454 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8N6G5 | |
Csgalnact2 | |
Synthetic peptide directed towards the C-terminal of human CSGALNACT2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |